Assay Details
Target Gene Details
Entrez Gene ID: | 21350 |
Gene Name: | T cell acute lymphocytic leukemia 2 |
Gene Aliases: |
bHLHa19 |
Location: |
Chr.4:53779705-53786885 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tal2 | NM_009317.3 | 2 | 783 | NP_033343.1 |
| AK018448.1 | 2 | 784 | BAB31216.1 | |
| BC110625.2 | 2 | 753 | AAI10626.1 |
More Information
Set Membership: |
|

Genomic Map