Assay Details
Target Gene Details
Entrez Gene ID: | 14581 |
Gene Name: | growth factor independent 1 |
Gene Aliases: |
AW495828, Gfi-1, Pal-1, Pal1 |
Location: |
Chr.5:107716655-107726031 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gfi1 | NM_001267621.1 | 7 | 1605 | NP_001254550.1 |
| NM_010278.2 | 6 | 1817 | NP_034408.1 | |
| AK089065.1 | 7 | 1606 | BAC40731.1 | |
| AK132814.1 | 1 | 473 | BAE21374.1 | |
| BC130219.1 | 5 | 1002 | ||
| BC152328.1 | 5 | 893 | ||
| BU698904.1 | ||||
| U58972.1 | 6 | 1792 | AAC52959.1 | |
| U78312.1 | 6 | 1817 | AAB36829.1 |
More Information
Set Membership: |
|

Genomic Map