Assay Details
Target Gene Details
Entrez Gene ID: | 212390 |
Gene Name: | kelch-like 32 |
Gene Aliases: |
6430524H05Rik, D4Ertd389e, Gm1356, mKIAA1900 |
Location: |
Chr.4:24614352-24851110 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Klhl32 | NM_001033531.3 | 6 | 1304 | NP_001028703.2 |
| NM_001163020.1 | 7 | 1403 | NP_001156492.1 | |
| AK134944.1 | 6 | 1276 | BAE22350.1 | |
| AK139083.1 | 2 | 645 | ||
| BC141239.1 | 6 | 1008 |

Genomic Map