Assay Details
Target Gene Details
Entrez Gene ID: | 242418 |
Gene Name: | DDB1 and CUL4 associated factor 10 |
Gene Aliases: |
AA959934, Wdr32 |
Location: |
Chr.4:45341735-45379732 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dcaf10 | NM_153167.2 | 1 | 315 | NP_694807.2 |
| AK045241.1 | 1 | 234 | BAC32276.1 | |
| AK157345.1 | 1 | 249 | ||
| AK165246.1 | 1 | 315 | BAE38101.1 | |
| AK170534.1 | 1 | 320 | ||
| BC027317.1 | 1 | 275 | AAH27317.1 | |
| BC085101.1 | 1 | 172 | AAH85101.1 | |
| BY732400.1 | 1 | 315 |

Genomic Map