Assay Details
Target Gene Details
Entrez Gene ID: | 242418 |
Gene Name: | DDB1 and CUL4 associated factor 10 |
Gene Aliases: |
AA959934, Wdr32 |
Location: |
Chr.4:45341735-45379732 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dcaf10 | NM_153167.2 | 7 | 3336 | NP_694807.2 |
| AK047592.1 | 1 | 1110 | ||
| AK076417.1 | 2 | 2096 | BAC36330.1 |

Genomic Map