Assay Details
Target Gene Details
Entrez Gene ID: | 68970 |
Gene Name: | DDB1 and CUL4 associated factor 12 |
Gene Aliases: |
1500001L20Rik, 5830424K06Rik, AA420338, AI851081, Wdr40a |
Location: |
Chr.4:41291300-41316726 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dcaf12 | NM_026893.3 | 1 | 305 | NP_081169.2 |
| AK005099.1 | 1 | 251 | BAB23817.2 | |
| AK030173.1 | 1 | 257 | BAC26822.1 | |
| AK031089.1 | 1 | 235 | BAC27247.1 | |
| AK083369.1 | 1 | 233 | ||
| AK087517.1 | 1 | 188 | BAC39908.1 | |
| AK090028.1 | 1 | 254 | BAC41053.1 | |
| AK143635.1 | 1 | 256 | ||
| AK173289.1 | 1 | 165 | BAD32567.1 | |
| BC023852.1 | 1 | 241 | AAH23852.1 | |
| BY338453.1 |

Genomic Map