Assay Details
Target Gene Details
Entrez Gene ID: | 231991 |
Gene Name: | cAMP responsive element binding protein 5 |
Gene Aliases: |
Crebpa, D430026C09Rik |
Location: |
Chr.6:53573374-53697850 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Creb5 | NM_172728.2 | 7 | 2551 | NP_766316.1 |
| AK051306.1 | ||||
| AK052451.1 | 7 | 2552 | BAC34998.1 | |
| AK154148.1 | 7 | 2552 | BAE32408.1 | |
| AK155537.1 | 7 | 2549 | BAE33314.1 | |
| BC029022.1 | 6 | 2486 | AAH29022.1 | |
| BC098487.1 | 7 | 2541 |
More Information
Set Membership: |
|

Genomic Map