Assay Details
Target Gene Details
Entrez Gene ID: | 171508 |
Gene Name: | cysteine-rich with EGF-like domains 1 |
Gene Aliases: |
AI843811 |
Location: |
Chr.6:113483297-113493338 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Creld1 | NM_133930.1 | 1 | 420 | NP_598691.1 |
| AB009020.1 | 1 | 425 | ||
| AK042344.1 | 1 | 567 | ||
| AK077726.1 | 1 | 564 | ||
| AK155777.1 | 1 | 430 | BAE33433.1 | |
| AK195695.1 | 1 | 106 | ||
| AK208544.1 | ||||
| BC010804.1 | 1 | 420 | AAH10804.1 | |
| BC023893.1 | 1 | 431 | AAH23893.1 | |
| BC025932.1 | 1 | 326 | AAH25932.1 | |
| BC029065.1 | 1 | 456 | AAH29065.1 |
More Information
Set Membership: |
|

Genomic Map