Assay Details
Target Gene Details
Entrez Gene ID: | 171508 |
Gene Name: | cysteine-rich with EGF-like domains 1 |
Gene Aliases: |
AI843811 |
Location: |
Chr.6:113483297-113493338 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Creld1 | NM_133930.1 | 10 | 1602 | NP_598691.1 |
| AB009020.1 | 10 | 1607 | ||
| AK050160.1 | 3 | 383 | BAC34101.1 | |
| AK050455.1 | 3 | 383 | BAC34266.1 | |
| AK077726.1 | 10 | 1747 | ||
| AK155777.1 | 10 | 1612 | BAE33433.1 | |
| AK186101.1 | ||||
| AK188085.1 | ||||
| AK213383.1 | ||||
| BC010804.1 | 10 | 1602 | AAH10804.1 | |
| BC023893.1 | 10 | 1613 | AAH23893.1 | |
| BC025932.1 | 10 | 1508 | AAH25932.1 | |
| BC029065.1 | 10 | 1638 | AAH29065.1 |
More Information
Set Membership: |
|

Genomic Map