Assay Details
Target Gene Details
Entrez Gene ID: | 26910 |
Gene Name: | folliculogenesis specific basic helix-loop-helix |
Gene Aliases: |
bHLHc8 |
Location: |
Chr.6:86010320-86020996 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Figla | NM_012013.1 | 1 | 84 | NP_036143.1 |
| JN700518.1 | 1 | 44 | ||
| U91840.1 | 1 | 84 | AAB97139.1 |
More Information
Set Membership: |
|

Genomic Map