Assay Details
Target Gene Details
Entrez Gene ID: | 50908 |
Gene Name: | complement component 1, s subcomponent 1 |
Gene Aliases: |
AA959438, AI255193, AI327365, C1s, C1sa |
Location: |
Chr.6:124530344-124542390 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C1s1 | NM_001097617.1 | 12 | 1756 | NP_001091086.1 |
| NM_144938.2 | 12 | 1793 | NP_659187.2 | |
| AF459019.1 | 12 | 1720 | AAO15558.1 | |
| AK087522.1 | 12 | 1725 | BAC39910.1 | |
| AK172453.1 | 12 | 1721 | BAE43013.1 | |
| BC018319.1 | 12 | 1685 | AAH18319.1 | |
| BC022123.1 | 12 | 1746 | AAH22123.1 |
More Information
Set Membership: |
|

Genomic Map