Assay Details
Target Gene Details
Entrez Gene ID: | 231876 |
Gene Name: | lemur tyrosine kinase 2 |
Gene Aliases: |
2900041G10Rik, A330101P12Rik, BREK, KPI-2, KPI2, aatyk2, cprk |
Location: |
Chr.5:144100366-144188204 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lmtk2 | NM_001081109.1 | 1 | 105 | NP_001074578.1 |
| AK158724.1 | 1 | 106 | BAE34627.1 |
More Information
Set Membership: |
|

Genomic Map