Assay Details
Target Gene Details
Entrez Gene ID: | 19301 |
Gene Name: | peroxisomal membrane protein 2 |
Gene Aliases: |
22kDa, PMP22 |
Location: |
Chr.5:110274286-110286168 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pxmp2 | NM_008993.2 | 1 | 240 | NP_033019.2 |
| AF309644.1 | 1 | 94 | AAG25724.1 | |
| AK003118.1 | 1 | 94 | BAB22578.1 | |
| BC057975.1 | 1 | 81 | AAH57975.1 | |
| BY090564.1 | 1 | 240 |

Genomic Map