Assay Details
Target Gene Details
Entrez Gene ID: | 319875 |
Gene Name: | transmembrane protease, serine 11B |
Gene Aliases: |
9930019B18Rik, Tmprss11bnl |
Location: |
Chr.5:86659522-86676298 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmprss11b | NM_177024.4 | 8 | 877 | NP_795998.2 |
| AK036968.1 | 8 | 930 | BAC29652.1 | |
| BC115419.1 | 8 | 877 | ||
| BC115420.2 | 8 | 877 |
More Information
Set Membership: |
|

Genomic Map