Assay Details
Target Gene Details
Entrez Gene ID: | 11517 |
Gene Name: | adenylate cyclase activating polypeptide 1 receptor 1 |
Gene Aliases: |
2900024I10Rik, AI846590, PAC1, PAC1R, PACAP1-R |
Location: |
Chr.6:55451501-55501455 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adcyap1r1 | NM_001025372.2 | 1 | 124 | NP_001020543.1 |
| NM_007407.4 | 1 | 124 | NP_031433.3 | |
| AK042820.1 | 1 | 116 | BAC31372.1 | |
| AK083108.1 | 1 | 122 | ||
| AK147291.1 | 1 | 75 | BAE27825.1 | |
| BC067039.1 | 1 | 57 | AAH67039.1 | |
| BY286680.1 | 1 | 125 |
More Information
Set Membership: |
|

Genomic Map