Assay Details
Target Gene Details
Entrez Gene ID: | 11517 |
Gene Name: | adenylate cyclase activating polypeptide 1 receptor 1 |
Gene Aliases: |
2900024I10Rik, AI846590, PAC1, PAC1R, PACAP1-R |
Location: |
Chr.6:55451501-55501455 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adcyap1r1 | NM_001025372.2 | 16 | 2434 | NP_001020543.1 |
| NM_007407.4 | 17 | 2518 | NP_031433.3 | |
| AK042820.1 | 17 | 2510 | BAC31372.1 | |
| AK047304.1 | 15 | 1937 | BAC33020.1 | |
| AK081121.1 | 15 | 1937 | BAC38140.1 | |
| AK083108.1 | 17 | 2517 | ||
| AK147291.1 | 16 | 2385 | BAE27825.1 | |
| AK147609.1 | 16 | 2175 | BAE28023.1 | |
| BC067039.1 | 16 | 2367 | AAH67039.1 |
More Information
Set Membership: |
|

Genomic Map