Assay Details
Target Gene Details
Entrez Gene ID: | 78600 |
Gene Name: | phosphodiesterase 6H, cGMP-specific, cone, gamma |
Gene Aliases: |
A930033D18Rik, PDEgamma |
Location: |
Chr.6:136952221-136963485 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pde6h | NM_023898.4 | 4 | 416 | NP_076387.2 |
| AK020922.1 | 4 | 417 | BAB32255.1 | |
| AK080696.1 | 4 | 432 | ||
| BC115671.1 | 4 | 394 | ||
| BC115672.1 | 4 | 394 | ||
| BE983779.1 | 1 | 159 |
More Information
Set Membership: |
|

Genomic Map