Assay Details
Target Gene Details
Entrez Gene ID: | 107999 |
Gene Name: | GTP binding protein 6 (putative) |
Gene Aliases: |
AV119224, Pgbpll, pgpl |
Location: |
Chr.5:110103976-110108197 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gtpbp6 | NM_145147.5 | 1 | 229 | NP_660129.2 |
| AK144812.1 | 1 | 230 | BAE26076.1 | |
| AK152225.1 | 1 | 230 | BAE31052.1 | |
| AK152972.1 | 1 | 230 | BAE31629.1 | |
| AK159462.1 | 1 | 202 | BAE35103.1 |

Genomic Map