Assay Details
Target Gene Details
Entrez Gene ID: | 22784 |
Gene Name: | solute carrier family 30 (zinc transporter), member 3 |
Gene Aliases: |
Znt3 |
Location: |
Chr.5:31086106-31108244 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc30a3 | NM_011773.3 | 8 | 1459 | NP_035903.2 |
| AK144568.1 | 8 | 1443 | BAE25941.1 | |
| AK165796.1 | 8 | 1422 | BAE38384.1 | |
| BC066199.1 | 8 | 1441 | AAH66199.1 | |
| U76007.1 | 8 | 1417 | AAB39731.1 |
More Information
Set Membership: |
|

Genomic Map