Assay Details
Target Gene Details
Entrez Gene ID: | 243084 |
Gene Name: | transmembrane protease, serine 11e |
Gene Aliases: |
A030012E10, AY787860, DESC1, TTSP |
Location: |
Chr.5:86705186-86745807 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmprss11e | NM_172880.2 | 8 | 811 | NP_766468.1 |
| AK037235.1 | 8 | 813 | BAC29770.1 | |
| AK162701.1 | 8 | 809 | ||
| AY787860.1 | 8 | 819 | AAV52922.1 | |
| BC115432.1 | 8 | 805 | ||
| BC115433.1 | 8 | 805 |
More Information
Set Membership: |
|

Genomic Map