Assay Details
Target Gene Details
Entrez Gene ID: | 22628 |
Gene Name: | tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, gamma polypeptide |
Gene Aliases: |
14-3-3gamma, D7Bwg1348e |
Location: |
Chr.5:135908379-135934641 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ywhag | NM_018871.3 | 1 | 213 | NP_061359.2 |
| AB073970.1 | 1 | 160 | ||
| AF058799.1 | 1 | 163 | AAC14345.1 | |
| AK076615.1 | 1 | 170 | BAC36417.1 | |
| AK088847.1 | 1 | 126 | BAC40609.1 | |
| AK148618.1 | 1 | 172 | BAE28625.1 | |
| AK153307.1 | 1 | 172 | BAE31888.1 | |
| AK164356.1 | 1 | 164 | BAE37756.1 | |
| BC008129.1 | 1 | 96 | AAH08129.1 | |
| BY244024.1 | 1 | 228 | ||
| CT010208.1 | CAJ18416.1 |
More Information
Set Membership: |
|

Genomic Map