Assay Details
Target Gene Details
Entrez Gene ID: | 170676 |
Gene Name: | paternally expressed 10 |
Gene Aliases: |
AA407948, Edr, HB-1, MEF3L, Mar2, Mart2, MyEF-3 |
Location: |
Chr.6:4747306-4760517 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Peg10 | NM_001040611.1 | 2 | 3460 | NP_001035701.1 |
| NM_130877.2 | 2 | 3460 | NP_570947.2 | |
| AB091827.1 | 3 | 3242 | BAC77244.1 | |
| AJ006464.1 | 3 | 3307 | ||
| CF735227.1 | 1 | 41 |

Genomic Map