Assay Details
Target Gene Details
Entrez Gene ID: | 74729 |
Gene Name: | SET domain without mariner transposase fusion |
Gene Aliases: |
5830404F24Rik, Etet2 |
Location: |
Chr.6:108065045-108077127 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Setmar | NM_001276356.1 | 3 | 820 | NP_001263285.1 |
| NM_178391.4 | 2 | 667 | NP_848478.2 | |
| AK017895.1 | 2 | 667 | ||
| BC045208.1 | 2 | 657 | AAH45208.1 |
More Information
Set Membership: |
|

Genomic Map