Assay Details
Target Gene Details
Entrez Gene ID: | 20388 |
Gene Name: | surfactant associated protein B |
Gene Aliases: |
AI562151, SF-B, SP-B, Sftp-3, Sftp3 |
Location: |
Chr.6:72304610-72314373 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sftpb | NM_001282071.2 | 11 | 1426 | NP_001269000.1 |
| NM_147779.2 | 11 | 1498 | NP_680088.1 |
More Information
Set Membership: |
|

Genomic Map