Assay Details
Target Gene Details
Entrez Gene ID: | 320737 |
Gene Name: | RIKEN cDNA 4732416N19 gene |
Gene Aliases: |
- |
Location: |
Chr.6:148212288-148235663 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4732416N19Rik | NR_015615.1 | |||
| AK005757.1 |
Target Gene Details
Entrez Gene ID: | 387314 |
Gene Name: | transmembrane and tetratricopeptide repeat containing 1 |
Gene Aliases: |
Arg99, BC023818 |
Location: |
Chr.6:148232430-148444845 on Build GRCm38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmtc1 | NM_198967.5 | 20 | 5596 | NP_945318.2 |
| AK135908.1 | 1 | 1011 |
More Information
Set Membership: |
|

Genomic Map