Assay Details
Target Gene Details
Entrez Gene ID: | 13448 |
Gene Name: | docking protein 1 |
Gene Aliases: |
AW557123, p62DOK |
Location: |
Chr.6:83030932-83033471 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dok1 | NM_001291799.1 | 5 | 1054 | NP_001278728.1 |
| NM_010070.4 | 5 | 1056 | NP_034200.4 | |
| AA790529.1 | ||||
| AK136382.1 | 5 | 1057 | BAE22956.1 | |
| AK169871.1 | 4 | 1222 | ||
| BC013066.1 | 5 | 1050 | AAH13066.1 | |
| U78818.2 | 5 | 1021 | AAB48827.2 |

Genomic Map