Assay Details
Target Gene Details
Entrez Gene ID: | 22044 |
Gene Name: | thyrotropin releasing hormone |
Gene Aliases: |
Pro-TRH, Trf |
Location: |
Chr.6:92242061-92244650 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trh | NM_009426.3 | 3 | 673 | NP_033452.2 |
| AK010666.1 | 3 | 672 | ||
| BC053493.1 | 3 | 659 | AAH53493.1 | |
| CF951860.1 | ||||
| X59387.1 | 2 | 623 | CAA42030.1 |
More Information
Set Membership: |
|

Genomic Map