Assay Details
Target Gene Details
Entrez Gene ID: | 64704 |
Gene Name: | HtrA serine peptidase 2 |
Gene Aliases: |
AI481710, Omi, Prss25, mnd2 |
Location: |
Chr.6:83051266-83054571 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Htra2 | NM_019752.3 | 8 | 1602 | NP_062726.3 |
| AF164513.1 | 8 | 1481 | AAF89534.1 | |
| AF175324.1 | 8 | 1498 | AAD50499.1 | |
| AK146338.1 | 8 | 1493 | BAE27092.1 | |
| AK159188.1 | 8 | 1422 | BAE34885.1 | |
| BC049880.1 | 8 | 1602 | AAH49880.1 | |
| BM899329.1 | 1 | 138 |
Target Gene Details
Entrez Gene ID: | 16950 |
Gene Name: | lysyl oxidase-like 3 |
Gene Aliases: |
Lor2, Loxl2 |
Location: |
Chr.6:83034224-83052564 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Loxl3 | NM_013586.4 | 14 | 2874 | NP_038614.2 |
| AK030548.1 | 14 | 2875 | BAC27016.1 | |
| AK145189.1 | 9 | 2048 | BAE26282.1 |
More Information
Set Membership: |
|

Genomic Map