Assay Details
Target Gene Details
Entrez Gene ID: | 19699 |
Gene Name: | reelin |
Gene Aliases: |
reeler, rl |
Location: |
Chr.5:21884454-22344705 on Build GRCm38 |
Assay Gene Location: | Within Exon 44 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Reln | NM_011261.2 | 44 | 7013 | NP_035391.2 |
| BC118041.1 | ||||
| D63520.1 | 3 | 263 | BAA09788.1 | |
| U24703.1 | 44 | 6998 | AAB91599.1 |
More Information
Set Membership: |
|

Genomic Map