Assay Details
Target Gene Details
Entrez Gene ID: | 12525 |
Gene Name: | CD8 antigen, alpha chain |
Gene Aliases: |
BB154331, Ly-2, Ly-35, Ly-B, Lyt-2 |
Location: |
Chr.6:71352772-71379171 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cd8a | NM_001081110.2 | 5 | 2736 | NP_001074579.1 |
| NM_009857.1 | 4 | 2705 | NP_033987.1 | |
| AJ131778.1 | 1 | 505 | ||
| AK037429.1 | 2 | 1042 | ||
| AK037467.1 | 5 | 2561 | BAC29812.1 | |
| AK088414.1 | 4 | 2580 | BAC40342.1 | |
| AK137937.1 | 6 | 2564 | BAE23516.1 |

Genomic Map