Assay Details
Target Gene Details
Entrez Gene ID: | 12576 |
Gene Name: | cyclin-dependent kinase inhibitor 1B |
Gene Aliases: |
AA408329, AI843786, Kip1, p27, p27Kip1 |
Location: |
Chr.6:134920401-134925525 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdkn1b | NM_009875.4 | 3 | 1341 | NP_034005.2 |
| AK162146.1 | 3 | 1061 |
More Information
Set Membership: |
|

Genomic Map