Assay Details
Target Gene Details
Entrez Gene ID: | 317677 |
Gene Name: | complement component 1, s subcomponent 2 |
Gene Aliases: |
C1sb, EG317677, Gm5077 |
Location: |
Chr.6:124624625-124636085 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C1s2 | NM_173864.2 | 11 | 1548 | NP_776289.2 |
| AF459020.1 | 11 | 1548 | AAO15559.1 |
More Information
Set Membership: |
|

Genomic Map