Assay Details
Target Gene Details
Entrez Gene ID: | 17295 |
Gene Name: | met proto-oncogene |
Gene Aliases: |
AI838057, HGF, HGFR, Par4, c-Met |
Location: |
Chr.6:17463349-17573980 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Met | NM_008591.2 | 1 | 199 | NP_032617.2 |
| AK148258.1 | 1 | 50 | ||
| AK170560.1 | 1 | 199 | BAE41881.1 | |
| CJ131939.1 | 1 | 356 |

Genomic Map