Assay Details
Target Gene Details
Entrez Gene ID: | 17295 |
Gene Name: | met proto-oncogene |
Gene Aliases: |
AI838057, HGF, HGFR, Par4, c-Met |
Location: |
Chr.6:17463349-17573980 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Met | NM_008591.2 | 3 | 545 | NP_032617.2 |
| AK148258.1 | 3 | 396 | ||
| AK170560.1 | 3 | 545 | BAE41881.1 | |
| BC117969.1 | 1 | 189 | ||
| BC117970.1 | 1 | 185 | ||
| BC128482.1 | ||||
| CJ131939.1 | ||||
| Y00671.1 | 1 | 190 | CAA68680.1 |

Genomic Map