Assay Details
Target Gene Details
Entrez Gene ID: | 66999 |
Gene Name: | mediator complex subunit 28 |
Gene Aliases: |
1500003D12Rik, AI451633, AU045690, Eg1, FKSG20, magicin |
Location: |
Chr.5:45520229-45529284 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Med28 | NM_025895.4 | 1 | 212 | NP_080171.1 |
| AF318060.1 | 1 | 136 | AAL26907.1 | |
| AK003854.1 | 1 | 125 | BAB23039.1 | |
| AK005681.1 | 1 | 121 | BAB24184.1 | |
| AK007855.1 | 1 | 141 | BAB25307.1 | |
| AK154249.1 | 1 | 138 | BAE32464.1 | |
| AK201969.1 | 1 | 53 | ||
| BC024941.1 | 1 | 155 | AAH24941.1 | |
| BY725533.1 | 1 | 192 | ||
| DV653139.1 | 1 | 212 |
More Information
Set Membership: |
|

Genomic Map