Assay Details
Target Gene Details
Entrez Gene ID: | 13848 |
Gene Name: | Eph receptor B6 |
Gene Aliases: |
Cekl, Mep |
Location: |
Chr.6:41605040-41620507 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ephb6 | NM_001146351.1 | 18 | 3658 | NP_001139823.1 |
| NM_007680.4 | 18 | 3654 | NP_031706.3 | |
| AK076442.1 | ||||
| AK087423.1 | 18 | 3569 | BAC39868.1 | |
| AK163823.1 | 9 | 1376 | BAE37507.1 | |
| BB833527.1 | 2 | 358 | ||
| BC031924.1 | 17 | 3266 | AAH31924.1 | |
| L77867.1 | 18 | 3348 | AAB51430.1 |
More Information
Set Membership: |
|

Genomic Map