Assay Details
Target Gene Details
Entrez Gene ID: | 242860 |
Gene Name: | round spermatid basic protein 1-like |
Gene Aliases: |
8430412F05Rik, AI447441, C330002G24Rik |
Location: |
Chr.5:20893024-20953441 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rsbn1l | NM_001080977.1 | 3 | 1207 | NP_001074446.1 |
| AK033374.1 | 3 | 887 |

Genomic Map