Assay Details
Target Gene Details
Entrez Gene ID: | 70118 |
Gene Name: | SRR1 domain containing |
Gene Aliases: |
2810002G02Rik, Srr1 |
Location: |
Chr.5:112337391-112343115 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Srrd | NM_027323.2 | 7 | 994 | NP_081599.2 |
| AK012640.1 | 7 | 959 | BAB28374.1 | |
| AK185989.1 | ||||
| AK188158.1 | ||||
| BC030678.1 | 5 | 603 | AAH30678.1 |
Target Gene Details
Entrez Gene ID: | 54723 |
Gene Name: | tuftelin interacting protein 11 |
Gene Aliases: |
2810002G02Rik, AF097181, AW046167, Srr1, TIP33, Tip39 |
Location: |
Chr.5:112326369-112338073 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tfip11 | NM_018783.4 | 14 | 2984 | NP_061253.2 |
| AF290474.3 | 14 | 2984 | AAG10198.2 | |
| AK148182.1 | 14 | 2989 | ||
| AK148260.1 | BAE28444.1 | |||
| AK211698.1 | ||||
| BC017682.1 | 14 | 2919 | AAH17682.1 |
More Information
Set Membership: |
|

Genomic Map