Assay Details
Target Gene Details
Entrez Gene ID: | 11921 |
Gene Name: | atonal homolog 1 (Drosophila) |
Gene Aliases: |
Hath1, MATH-1, Math1, bHLHa14 |
Location: |
Chr.6:64729146-64731243 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atoh1 | NM_007500.4 | 1 | 1395 | NP_031526.1 |
| AK082354.1 | 1 | 1417 | ||
| BC010820.1 | 1 | 1395 | AAH10820.1 | |
| BC051256.1 | 1 | 1412 | AAH51256.1 |
More Information
Set Membership: |
|

Genomic Map