Assay Details
Target Gene Details
Entrez Gene ID: | 76252 |
Gene Name: | ATPase, H+ transporting, lysosomal V0 subunit E2 |
Gene Aliases: |
0610006O14Rik, NM9.2 |
Location: |
Chr.6:48537569-48541801 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atp6v0e2 | NM_133764.3 | 4 | 921 | NP_598525.1 |
| AK002260.1 | 4 | 789 | ||
| AK077313.1 | 4 | 793 | ||
| BC010790.1 | 4 | 875 | AAH10790.1 |
More Information
Set Membership: |
|

Genomic Map