Assay Details
Target Gene Details
Entrez Gene ID: | 232717 |
Gene Name: | protease, serine 58 |
Gene Aliases: |
1700020H15, Tryx3 |
Location: |
Chr.6:40895262-40900387 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prss58 | NM_175020.3 | 4 | 436 | NP_778185.1 |
| AK075754.1 | 4 | 437 | BAC35932.1 | |
| BC048599.1 | 4 | 430 | AAH48599.1 |
More Information
Set Membership: |
|

Genomic Map