Assay Details
Target Gene Details
Entrez Gene ID: | 73998 |
Gene Name: | hect domain and RLD 3 |
Gene Aliases: |
5730409F18Rik, AI646603, mKIAA0032 |
Location: |
Chr.6:58831477-58920398 on Build GRCm38 |
Assay Gene Location: | Within Intron 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Herc3 | NM_028705.3 | NP_082981.3 | ||
| AK122192.1 | BAC65474.3 | |||
| BC042574.1 | AAH42574.1 | |||
| BC145978.1 | ||||
| BC145980.1 |
Target Gene Details
Entrez Gene ID: | 58243 |
Gene Name: | nucleosome assembly protein 1-like 5 |
Gene Aliases: |
1110020M21Rik |
Location: |
Chr.6:58905233-58907126 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nap1l5 | NM_021432.2 | 1 | 634 | NP_067407.1 |
| AB041556.1 | 1 | 573 | BAA95041.1 | |
| AK003875.1 | 1 | 340 | BAB23053.1 | |
| BC046332.1 | 1 | 584 | AAH46332.1 | |
| BU526293.1 | 1 | 637 |
More Information
Set Membership: |
|

Genomic Map