Assay Details
Target Gene Details
Entrez Gene ID: | 20893 |
Gene Name: | basic helix-loop-helix family, member e40 |
Gene Aliases: |
Bhlhb2, C130042M06Rik, CR8, Clast5, Dec1, Sharp2, Stra13, Stra14 |
Location: |
Chr.6:108660629-108666925 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bhlhe40 | NM_011498.4 | 5 | 1888 | NP_035628.1 |
| AF010305.1 | 5 | 1820 | AAB64228.1 | |
| AF364051.1 | 5 | 1820 | AAK50859.1 | |
| AK048242.1 | 5 | 1889 | BAC33281.1 | |
| AK090032.1 | 5 | 1842 | BAC41056.1 | |
| AK154935.1 | 5 | 1841 | BAE32936.1 | |
| BC010720.1 | 5 | 1831 | AAH10720.1 | |
| BC094057.1 | 1 | 943 | AAH94057.1 |
More Information
Set Membership: |
|

Genomic Map