Assay Details
Target Gene Details
Entrez Gene ID: | 387353 |
Gene Name: | taste receptor, type 2, member 126 |
Gene Aliases: |
T2R12, T2R26, Tas2r26, Tas2r41, mGR26, mt2r35 |
Location: |
Chr.6:42434535-42435461 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tas2r126 | NM_207028.1 | 1 | 309 | NP_996911.1 |
| BC126974.1 | 1 | 309 |
More Information
Set Membership: |
|

Genomic Map