Assay Details
Target Gene Details
Entrez Gene ID: | 73408 |
Gene Name: | RIKEN cDNA 1700065L07 gene |
Gene Aliases: |
- |
Location: |
Chr.6:73436965-73471021 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 1700065L07Rik | NR_108032.1 | |||
| AK006896.1 |
Target Gene Details
Entrez Gene ID: | 66740 |
Gene Name: | RIKEN cDNA 4931417E11 gene |
Gene Aliases: |
- |
Location: |
Chr.6:73468573-73469667 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4931417E11Rik | NM_025737.3 | 1 | 911 | NP_080013.1 |
| AK005631.1 | 1 | 836 | BAB24158.1 | |
| AK016460.1 | 1 | 912 | BAB30249.1 | |
| BB013990.1 | 1 | 68 | ||
| BC048456.1 | 1 | 863 | AAH48456.1 |
More Information
Set Membership: |
|

Genomic Map