Assay Details
Target Gene Details
Entrez Gene ID: | 20678 |
Gene Name: | SRY (sex determining region Y)-box 5 |
Gene Aliases: |
A730017D01Rik, AI528773 |
Location: |
Chr.6:143828425-144781989 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sox5 | AK029047.1 | |||
| AK087772.1 |
Target Gene Details
Entrez Gene ID: | 66760 |
Gene Name: | SRY (sex determining region Y)-box 5, opposite strand 3 |
Gene Aliases: |
4933425H06Rik |
Location: |
Chr.6:144672868-144693832 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sox5os3 | NR_040519.1 | |||
| AK006194.1 | ||||
| AK016914.1 |

Genomic Map