Assay Details
Target Gene Details
Entrez Gene ID: | 66964 |
Gene Name: | golgi transport 1 homolog B (S. cerevisiae) |
Gene Aliases: |
2310061A22Rik, AA407874, AU024291, CGI-141 |
Location: |
Chr.6:142387209-142410084 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Golt1b | NM_025872.4 | 5 | 1255 | NP_080148.1 |
| AK145964.1 | 3 | 992 | BAE26789.1 | |
| BC085246.1 | 5 | 1256 | AAH85246.1 |
More Information
Set Membership: |
|

Genomic Map