Assay Details
Target Gene Details
Entrez Gene ID: | 93695 |
Gene Name: | glycoprotein (transmembrane) nmb |
Gene Aliases: |
DC-HIL, Dchil, ipd |
Location: |
Chr.6:49036518-49058182 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gpnmb | NM_053110.4 | 6 | 1026 | NP_444340.3 |
| AF322054.1 | 6 | 864 | AAK14240.1 | |
| AJ251685.1 | 6 | 911 | CAB65272.1 | |
| AK044764.1 | 6 | 920 | BAC32074.1 | |
| AK076347.1 | 6 | 895 | BAC36306.1 | |
| AK149700.1 | BAE29036.1 | |||
| AK149740.1 | 6 | 933 | ||
| AK150739.1 | 6 | 895 | BAE29813.1 | |
| AK154617.1 | 6 | 930 | BAE32716.1 | |
| AK159165.1 | 6 | 925 | BAE34867.1 | |
| AK159270.1 | 6 | 933 | BAE34949.1 | |
| AK159747.1 | 6 | 895 | BAE35339.1 | |
| AK159908.1 | 6 | 901 | ||
| AK170506.1 | 6 | 925 | BAE41844.1 | |
| AK171347.1 | 6 | 901 | BAE42404.1 | |
| AK171441.1 | 6 | 895 | BAE42454.1 | |
| AK171465.1 | 6 | 895 | BAE42471.1 | |
| AK171474.1 | 6 | 895 | BAE42479.1 | |
| AK171517.1 | 6 | 1013 | BAE42501.1 | |
| AK171608.1 | 6 | 895 | BAE42560.1 | |
| AK171618.1 | 6 | 895 | BAE42567.1 | |
| AK179971.1 | 3 | 303 | ||
| AK192937.1 | 2 | 170 | ||
| AK193247.1 | 2 | 170 | ||
| AK193314.1 | 2 | 170 | ||
| AK193818.1 | 2 | 170 | ||
| AK195422.1 | 2 | 170 | ||
| AK196346.1 | 2 | 170 | ||
| AK196951.1 | 2 | 170 | ||
| AK201982.1 | 2 | 250 | ||
| AK204190.1 | 2 | 170 | ||
| AK214279.1 | 2 | 170 | ||
| AK215880.1 | 2 | 170 | ||
| BC026375.1 | 6 | 900 | AAH26375.1 |
More Information
Set Membership: |
|

Genomic Map