Assay Details
Target Gene Details
Entrez Gene ID: | 58887 |
Gene Name: | replication initiator 1 |
Gene Aliases: |
AI425994, Ap4, E430037F08Rik, Zfp-464, Zfp464 |
Location: |
Chr.6:48593883-48599082 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Repin1 | NM_001079901.1 | 3 | 2209 | NP_001073370.2 |
| NM_001079902.1 | 3 | 2206 | NP_001073371.2 | |
| NM_001079903.1 | 3 | 2138 | NP_001073372.2 | |
| NM_001079904.1 | 3 | 2135 | NP_001073373.2 | |
| NM_001079905.1 | 2 | 2020 | NP_001073374.1 | |
| NM_175099.3 | 2 | 2023 | NP_780308.2 | |
| AK089041.1 | 2 | 1967 | BAC40716.1 | |
| AK137310.1 | 3 | 2082 | ||
| AK170797.1 | 3 | 2174 | ||
| BC085128.1 | 3 | 2141 | AAH85128.1 |
More Information
Set Membership: |
|

Genomic Map