Assay Details
Target Gene Details
Entrez Gene ID: | 242864 |
Gene Name: | N-acyl phosphatidylethanolamine phospholipase D |
Gene Aliases: |
A530089G06, Mbldc1, NAPE-PLD |
Location: |
Chr.5:21662901-21702153 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Napepld | NM_178728.5 | 3 | 572 | NP_848843.1 |
| AB112350.1 | 2 | 415 | BAD02397.1 | |
| AK041194.1 | 3 | 570 | BAC30857.1 | |
| AK049135.1 | 3 | 831 | BAC33563.1 | |
| AK080888.1 | 3 | 572 | BAC38064.1 | |
| AK141585.1 | 3 | 852 | BAE24749.1 | |
| BC062890.1 | 3 | 594 | AAH62890.1 |
More Information
Set Membership: |
|

Genomic Map